Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chadl
Entrez Gene ID
100910434
Gene Name
chondroadherin-like
Gene Aliases
-
Chromosome Location
Chr.7: 122915847 - 122929114 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008765807.1 XP_008764029.1 1 - 2 307 80
XM_008765808.1 XP_008764030.1 1 - 2 307 80
XM_008765809.1 XP_008764031.1 2 - 3 1647 80

View Full Product Details