The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011990.1 | NP_001011990.1 | 12 - 13 | 1305 | 132 |
GenBank mRNA | BC082087.1 | - | 12 - 13 | 1305 | 132 |
FQ215133.1 | - | 10 - 11 | 1116 | 132 | |
FQ216598.1 | - | 11 - 12 | 1116 | 132 | |
FQ228717.1 | - | 11 - 12 | 1116 | 132 |