The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008384.1 | NP_001008385.1 | 4 - 5 | 381 | 57 |
XM_006242028.2 | XP_006242090.1 | 4 - 5 | 424 | 57 | |
GenBank mRNA | BC086399.1 | - | 4 - 5 | 381 | 57 |
FQ213630.1 | - | 4 - 5 | 380 | 57 | |
FQ225192.1 | - | 4 - 5 | 385 | 57 | |
FQ230979.1 | - | 4 - 5 | 423 | 57 | |
FQ232209.1 | - | 4 - 5 | 405 | 57 | |
FQ234815.1 | - | 4 - 5 | 405 | 57 | |
FQ234871.1 | - | 4 - 5 | 358 | 57 |