The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008384.1 | NP_001008385.1 | 6 - 7 | 681 | 69 |
GenBank mRNA | BC086399.1 | - | 6 - 7 | 681 | 69 |
FQ213630.1 | - | 6 - 7 | 680 | 69 | |
FQ225192.1 | - | 6 - 7 | 685 | 69 | |
FQ232209.1 | - | 6 - 7 | 705 | 69 | |
FQ234815.1 | - | 6 - 7 | 705 | 69 | |
FQ234871.1 | - | 6 - 7 | 658 | 69 |