Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ly6d
Entrez Gene ID
315075
Gene Name
lymphocyte antigen 6 complex, locus D
Gene Aliases
-
Chromosome Location
Chr.7: 115993304 - 115994812 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130552.1 NP_001124024.1 2 - 3 205 89

View Full Product Details