Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rrm2b
Entrez Gene ID
299976
Gene Name
ribonucleotide reductase M2 B (TP53 inducible)
Gene Aliases
-
Chromosome Location
Chr.7: 76750045 - 76780817 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130543.1 NP_001124015.1 1 - 2 179 82

View Full Product Details