Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp2d4
Entrez Gene ID
171522
Gene Name
cytochrome P450, family 2, subfamily d, polypeptide 4
Gene Aliases
Cyp2d18, Cyp2d22, Cyp2d4v1, Cyp2d4v2, Cyp2d6
Chromosome Location
Chr.7: 123599264 - 123608436 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138515.2 NP_612524.1 4 - 5 708 102
GenBank mRNA AB008425.1 - 3 - 4 581 102
BC093609.1 - - 723 102
FQ211703.1 - - 724 102
U48219.1 - 4 - 5 708 102
U48220.1 - - 706 102

View Full Product Details