Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trps1
Entrez Gene ID
299897
Gene Name
trichorhinophalangeal syndrome I
Gene Aliases
-
Chromosome Location
Chr.7: 90090828 - 90318748 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134837.1 NP_001128309.1 5 - 6 2851 99

View Full Product Details