Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Khdrbs3
Entrez Gene ID
64015
Gene Name
KH domain containing, RNA binding, signal transduction associated 3
Gene Aliases
Etle, Slm2
Chromosome Location
Chr.7: 110031819 - 110179475 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022249.1 NP_071585.1 5 - 6 726 65
GenBank mRNA AF152547.1 - 5 - 6 726 65

View Full Product Details