Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsta3
Entrez Gene ID
300036
Gene Name
tissue specific transplantation antigen P35B
Gene Aliases
-
Chromosome Location
Chr.7: 116958427 - 116963281 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127455.2 NP_001120927.1 2 - 3 192 67
NM_001270789.1 NP_001257718.1 2 - 3 258 67
NM_001270790.1 NP_001257719.1 2 - 3 258 67
GenBank mRNA BC158885.1 - 2 - 3 230 67
BF289545.1 - 2 - 3 193 67
CA339313.1 - 1 - 2 146 67
CK653514.1 - - 191 67
FQ210097.1 - 2 - 3 67
FQ221078.1 - 2 - 3 180 67
FQ229919.1 - 2 - 3 203 67

Gene Symbol
Pibf1
Entrez Gene ID
306104
Gene Name
progesterone immunomodulatory binding factor 1
Gene Aliases
RGD1305077
Chromosome Location
Chr.15: 83494146 - 83663202 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FQ210517.1 - - 67

View Full Product Details