Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcf20
Entrez Gene ID
366964
Gene Name
transcription factor 20
Gene Aliases
AR1, SPBP
Chromosome Location
Chr.7: 123670135 - 123767797 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130574.1 NP_001124046.1 3 - 4 6029 80
XM_006242110.2 XP_006242172.1 2 - 3 6929 80
XM_006242112.2 XP_006242174.1 2 - 3 6929 80
XM_008765772.1 XP_008763994.1 2 - 3 6928 80

View Full Product Details