Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Samd12
Entrez Gene ID
362910
Gene Name
sterile alpha motif domain containing 12
Gene Aliases
RGD1561402
Chromosome Location
Chr.7: 92995599 - 93286757 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130562.1 NP_001124034.1 2 - 3 247 143

View Full Product Details