Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gga1
Entrez Gene ID
300066
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 120102592 - 120119738 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011994.1 NP_001011994.1 3 - 4 396 98
XM_006241973.2 XP_006242035.1 4 - 5 1758 98
XM_008765679.1 XP_008763901.1 5 - 6 1514 98
XM_008765680.1 XP_008763902.1 2 - 3 1012 98
XM_008765681.1 XP_008763903.1 4 - 5 4220 98
GenBank mRNA BC090031.1 - 3 - 4 396 98

View Full Product Details