Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp2d2
Entrez Gene ID
25053
Gene Name
cytochrome P450, family 2, subfamily d, polypeptide 2
Gene Aliases
Cyp2d26
Chromosome Location
Chr.7: 123651827 - 123655896 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012730.1 NP_036862.1 1 - 2 317 89
GenBank mRNA AB008423.1 - 1 - 2 193 89
BC078897.1 - 1 - 2 252 89
FQ218496.1 - 1 - 2 258 89
M16655.1 - 1 - 2 243 89
M22330.1 - 1 - 2 317 89

View Full Product Details