Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp2d2
Entrez Gene ID
25053
Gene Name
cytochrome P450, family 2, subfamily d, polypeptide 2
Gene Aliases
Cyp2d26
Chromosome Location
Chr.7: 123651827 - 123655896 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB008423.1 - 2 - 3 351 82
BC078897.1 - 2 - 3 410 82
FQ218496.1 - 2 - 3 416 82
M16655.1 - 2 - 3 401 82

View Full Product Details