Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam91a1
Entrez Gene ID
689997
Gene Name
family with sequence similarity 91, member A1
Gene Aliases
-
Chromosome Location
Chr.7: 98302802 - 98341201 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127578.1 NP_001121050.1 21 - 22 2356 65
XM_006241684.2 XP_006241746.1 21 - 22 2360 65
GenBank mRNA BC166852.1 - 21 - 22 2356 65

View Full Product Details