The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033703.1 | NP_001028875.1 | 1 - 2 | 144 | 97 |
XM_008765716.1 | XP_008763938.1 | 1 - 2 | 1280 | 97 | |
GenBank mRNA | BC101878.1 | - | 1 - 2 | 144 | 97 |