Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atxn10
Entrez Gene ID
170821
Gene Name
ataxin 10
Gene Aliases
Sca10
Chromosome Location
Chr.7: 126228416 - 126351728 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133313.2 NP_579847.1 6 - 7 824 76
GenBank mRNA AJ301634.1 - 6 - 7 933 76
BC062087.1 - 6 - 7 824 76
FQ213088.1 - 1 - 2 56 76

View Full Product Details