Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syngr1
Entrez Gene ID
29205
Gene Name
synaptogyrin 1
Gene Aliases
-
Chromosome Location
Chr.7: 121311010 - 121334437 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006241971.2 XP_006242033.1 3 - 4 540 121
GenBank mRNA DN932005.1 - 3 - 4 497 121
DV724604.1 - 2 - 3 341 121

View Full Product Details