Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Upk3a
Entrez Gene ID
315190
Gene Name
uroplakin 3A
Gene Aliases
-
Chromosome Location
Chr.7: 125920357 - 125925430 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130507.1 NP_001123979.1 4 - 5 598 74

View Full Product Details