The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013949.1 | NP_001013971.2 | 6 - 7 | 1224 | 106 |
GenBank mRNA | BC079434.1 | - | 6 - 7 | 1224 | 106 |