Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Naprt1
Entrez Gene ID
315085
Gene Name
nicotinate phosphoribosyltransferase domain containing 1
Gene Aliases
Naprt
Chromosome Location
Chr.7: 116922979 - 116926436 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207609.1 NP_997492.1 6 - 7 958 63
GenBank mRNA AY214330.1 - 6 - 7 958 63

View Full Product Details