Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St3gal1
Entrez Gene ID
362924
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 1
Gene Aliases
Siat4a
Chromosome Location
Chr.7: 107895045 - 107963150 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013219.1 NP_001013237.1 3 - 4 685 136
XM_006241709.2 XP_006241771.1 6 - 7 1657 136
XM_006241710.2 XP_006241772.1 6 - 7 2575 136
XM_008765513.1 XP_008763735.1 5 - 6 1602 136
GenBank mRNA AJ748840.1 - 3 - 4 685 136

View Full Product Details