The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011991.1 | NP_001011991.1 | 15 - 16 | 1085 | 73 |
XM_006241708.2 | XP_006241770.1 | 15 - 16 | 1357 | 73 | |
GenBank mRNA | AY500369.1 | - | 15 - 16 | 1047 | 73 |
BC081898.1 | - | 15 - 16 | 1085 | 73 | |
FQ212154.1 | - | 15 - 16 | 1097 | 73 |