The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013104.1 | NP_001013122.1 | 7 - 8 | 1917 | 80 |
XM_008765549.1 | XP_008763771.1 | 8 - 9 | 4038 | 80 | |
XM_008765550.1 | XP_008763772.1 | 8 - 9 | 2099 | 80 | |
XM_008765551.1 | XP_008763773.1 | 10 - 11 | 2091 | 80 | |
XM_008765552.1 | XP_008763774.1 | 10 - 11 | 2072 | 80 | |
XM_008765553.1 | XP_008763775.1 | 8 - 9 | 1991 | 80 | |
XM_008765554.1 | XP_008763776.1 | 7 - 8 | 2045 | 80 | |
XM_008765555.1 | XP_008763777.1 | 7 - 8 | 2030 | 80 | |
XM_008765556.1 | XP_008763778.1 | 6 - 7 | 1973 | 80 | |
XM_008765557.1 | XP_008763779.1 | 7 - 8 | 921 | 80 | |
XM_008765558.1 | XP_008763780.1 | 7 - 8 | 928 | 80 | |
XM_008765559.1 | XP_008763781.1 | 7 - 8 | 908 | 80 | |
XM_008765560.1 | XP_008763782.1 | 7 - 8 | 906 | 80 | |
XM_008765561.1 | XP_008763783.1 | 6 - 7 | 849 | 80 | |
XM_008765562.1 | XP_008763784.1 | 6 - 7 | 834 | 80 | |
GenBank mRNA | AF145050.1 | - | 1 - 2 | 101 | 80 |
BC079391.1 | - | 7 - 8 | 1917 | 80 | |
FQ225933.1 | - | 7 - 8 | 913 | 80 | |
FQ227443.1 | - | 7 - 8 | 889 | 80 | |
FQ233760.1 | - | 7 - 8 | 869 | 80 |