The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007146.1 | NP_001007147.1 | 1 - 2 | 157 | 103 |
GenBank mRNA | AY724519.1 | - | 1 - 2 | 360 | 103 |
BC085117.1 | - | 1 - 2 | 196 | 103 |