The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113785.1 | NP_001107257.1 | 8 - 9 | 727 | 144 |
GenBank mRNA | BC158805.1 | - | 8 - 9 | 727 | 144 |
FQ214096.1 | - | 5 - 6 | 610 | 144 | |
FQ215024.1 | - | 8 - 9 | 716 | 144 |