Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cbx7
Entrez Gene ID
362962
Gene Name
chromobox homolog 7
Gene Aliases
-
Chromosome Location
Chr.7: 121136067 - 121153664 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199117.2 NP_954548.1 5 - 6 533 80
XM_006242020.2 XP_006242082.1 5 - 6 942 80
XM_006242021.2 XP_006242083.1 5 - 6 942 80
XM_006242024.2 XP_006242086.1 5 - 6 748 80
XM_006242025.2 XP_006242087.1 5 - 6 638 80
GenBank mRNA BC062392.1 - 5 - 6 513 80

View Full Product Details