Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Micalcl
Entrez Gene ID
293180
Gene Name
MICAL C-terminal like
Gene Aliases
RSB-11-77
Chromosome Location
Chr.1: 177193735 - 177232674 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182669.2 NP_872610.2 3 - 4 284 82
GenBank mRNA BC098647.1 - 3 - 4 284 82

View Full Product Details