Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gp2
Entrez Gene ID
171459
Gene Name
glycoprotein 2 (zymogen granule membrane)
Gene Aliases
gp80
Chromosome Location
Chr.1: 189166735 - 189182306 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134418.3 NP_599245.2 3 - 4 623 73
XM_008759672.1 XP_008757894.1 3 - 4 619 73
GenBank mRNA M58716.1 - 3 - 4 625 73
X53935.1 - 2 - 3 524 73

View Full Product Details