Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ric3
Entrez Gene ID
687147
Gene Name
RIC3 acetylcholine receptor chaperone
Gene Aliases
-
Chromosome Location
Chr.1: 173628281 - 173682274 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115045.3 NP_001108517.1 2 - 3 354 106
NM_001277069.1 NP_001263998.1 2 - 3 354 106
GenBank mRNA AM422212.1 - 2 - 3 354 106
AM422213.1 - 2 - 3 354 106

View Full Product Details