Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nlrp10
Entrez Gene ID
293426
Gene Name
NLR family, pyrin domain containing 10
Gene Aliases
Nalp10
Chromosome Location
Chr.1: 173521412 - 173528969 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106291.1 NP_001099761.1 2 - 3 367 125

View Full Product Details