The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011933.1 | NP_001011933.1 | 11 - 12 | 1451 | 115 |
XM_006230017.2 | XP_006230079.1 | 12 - 13 | 1533 | 115 | |
XM_006230018.2 | XP_006230080.1 | 12 - 13 | 1458 | 115 | |
XM_006230020.2 | XP_006230082.1 | 12 - 13 | 1535 | 115 | |
GenBank mRNA | BC082015.1 | - | 11 - 12 | 1451 | 115 |
FQ221385.1 | - | 12 - 13 | 1517 | 115 |