Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nlrp14
Entrez Gene ID
499234
Gene Name
NLR family, pyrin domain containing 14
Gene Aliases
-
Chromosome Location
Chr.1: 171305550 - 171340428 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008759845.1 XP_008758067.1 9 - 10 3256 124

View Full Product Details