Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Htra1
- Entrez Gene ID
-
65164
- Gene Name
- HtrA serine peptidase 1
- Gene Aliases
-
Prss11
- Chromosome Location
-
Chr.1: 201499067 - 201548508 on Build Rnor_6.0
- Gene Symbol
- Psmd1
- Entrez Gene ID
-
83806
- Gene Name
- proteasome (prosome, macropain) 26S subunit, non-ATPase, 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 93080618 - 93155033 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
FQ228875.1 |
- |
- |
93 |
68 |
View Full Product Details