Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lat
Entrez Gene ID
81511
Gene Name
linker for activation of T cells
Gene Aliases
-
Chromosome Location
Chr.1: 197765642 - 197771123 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030853.1 NP_110480.1 11 - 12 971 72
XM_008759919.1 XP_008758141.1 10 - 11 1395 72
GenBank mRNA AJ001184.1 - 11 - 12 971 72
BC088130.1 - 10 - 11 999 72
FQ233731.1 - 10 - 11 958 72

View Full Product Details