Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab6a
Entrez Gene ID
84379
Gene Name
RAB6A, member RAS oncogene family
Gene Aliases
RCO4-3, Rab6, Rab6b
Chromosome Location
Chr.1: 165624973 - 165664201 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053366.1 NP_445818.1 4 - 5 733 163

View Full Product Details