Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rgs10
Entrez Gene ID
54290
Gene Name
regulator of G-protein signaling 10
Gene Aliases
-
Chromosome Location
Chr.1: 199782076 - 199823439 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019337.1 NP_062210.1 1 - 2 96
XM_006230326.2 XP_006230388.1 1 - 2 118 96
GenBank mRNA CD372843.1 - 1 - 2 96

View Full Product Details