Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Insc
Entrez Gene ID
293166
Gene Name
inscuteable homolog (Drosophila)
Gene Aliases
RGD1311379
Chromosome Location
Chr.1: 184299250 - 184407480 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106285.1 NP_001099755.1 3 - 4 705 77

View Full Product Details