Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Setd1b
Entrez Gene ID
309001
Gene Name
SET domain containing 1B
Gene Aliases
RGD1311624
Chromosome Location
Chr.1: 199222586 - 199247993 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC098812.1 - 1 - 2 112 58

View Full Product Details