The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012137.1 | NP_001012137.1 | 6 - 7 | 807 | 137 |
XM_006230290.2 | XP_006230352.1 | 6 - 7 | 831 | 137 | |
GenBank mRNA | BC087153.1 | - | 6 - 7 | 807 | 137 |