Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vom2r40
Entrez Gene ID
286983
Gene Name
vomeronasal 2 receptor, 40
Gene Aliases
-
Chromosome Location
Chr.1: 148927532 - 148974509 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173317.1 NP_775439.1 - 1111 123
GenBank mRNA AF016180.1 - - 1111 123

View Full Product Details