Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Vom2r40
- Entrez Gene ID
-
286983
- Gene Name
- vomeronasal 2 receptor, 40
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 148927532 - 148974509 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_173317.1 |
NP_775439.1 |
- |
1924 |
118 |
GenBank mRNA |
AF016180.1 |
- |
- |
1924 |
118 |
- Gene Symbol
- Vom2r41
- Entrez Gene ID
-
293069
- Gene Name
- vomeronasal 2 receptor, 41
- Gene Aliases
-
RGD1561322
- Chromosome Location
-
-
View Full Product Details