Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bcl7c
Entrez Gene ID
293514
Gene Name
B-cell CLL/lymphoma 7C
Gene Aliases
-
Chromosome Location
Chr.1: 199155296 - 199159125 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106298.1 NP_001099768.1 3 - 4 546 107

View Full Product Details