Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sept1
Entrez Gene ID
293507
Gene Name
septin 1
Gene Aliases
-
Chromosome Location
Chr.1: 198658744 - 198662610 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012460.1 NP_001012478.1 8 - 9 793 128
GenBank mRNA BC089897.1 - 8 - 9 793 128

View Full Product Details