Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tufm
Entrez Gene ID
293481
Gene Name
Tu translation elongation factor, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.1: 197903582 - 197907189 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106295.1 NP_001099765.1 4 - 5 853 109
XM_006230224.1 XP_006230286.1 4 - 5 848 109
XM_006230225.1 XP_006230287.1 4 - 5 848 109

View Full Product Details