Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC100909966
Entrez Gene ID
100909966
Gene Name
olfactory receptor 6F1-like
Gene Aliases
-
Chromosome Location
Chr.1: 149602346 - 149603284 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003748916.2 XP_003748964.1 - 856 91

Gene Symbol
Olr13
Entrez Gene ID
293075
Gene Name
olfactory receptor 13
Gene Aliases
-
Chromosome Location
Chr.1: 149786382 - 149787308 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001000116.1 NP_001000116.1 1 - 1 844 91

View Full Product Details