Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cradd
Entrez Gene ID
314756
Gene Name
CASP2 and RIPK1 domain containing adaptor with death domain
Gene Aliases
RAIDD
Chromosome Location
Chr.7: 36395665 - 36408588 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108085.1 NP_001101555.1 2 - 3 457 121

View Full Product Details