Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Poc1b
Entrez Gene ID
100362836
Gene Name
POC1 centriolar protein B
Gene Aliases
-
Chromosome Location
Chr.7: 41304418 - 41403483 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729781.3 XP_002729827.2 2 - 3 180 88
XM_008765341.1 XP_008763563.1 2 - 3 345 88
GenBank mRNA FQ231893.1 - 2 - 3 244 88
FQ235303.1 - 2 - 3 244 88

View Full Product Details