Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Glipr1l2
Entrez Gene ID
366890
Gene Name
GLI pathogenesis-related 1 like 2
Gene Aliases
RGD1310205
Chromosome Location
Chr.7: 54795174 - 54824255 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729783.4 XP_002729829.3 1 - 2 569 75

View Full Product Details